Assay Details
Target Gene Details
Entrez Gene ID: | 92345 |
Gene Name: | nuclear assembly factor 1 ribonucleoprotein |
Gene Aliases: |
- |
Location: |
Chr.4:163109133-163170388 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAF1 | AK126037.1 | 1 | 1604 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015505 | Chr.4:161989825 - 163516137 on Build GRCh38 | Gain |
|
| nsv1030184 | Chr.4:162402427 - 164455529 on Build GRCh38 | Loss |
|
| nsv4584 | Chr.4:163091996 - 163125621 on Build GRCh38 | Insertion |
|
| esv3894064 | Chr.4:162963457 - 163303980 on Build GRCh38 | Gain |
|

Genomic Map