Assay Details
Target Gene Details
Entrez Gene ID: | 90632 |
Gene Name: | long intergenic non-protein coding RNA 473 |
Gene Aliases: |
C6orf176, bA142J11.1 |
Location: |
Chr.6:165924048-165988039 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00473 | NR_026861.1 | |||
| CB270066.1 | ||||
| DA416538.1 |
Target Gene Details
Entrez Gene ID: | 10846 |
Gene Name: | phosphodiesterase 10A |
Gene Aliases: |
ADSD2, HSPDE10A, IOLOD, LINC00473, PDE10A19 |
Location: |
Chr.6:165327287-165988121 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE10A | XM_011535387.2 | XP_011533689.2 | ||
| XM_017010194.1 | XP_016865683.1 | |||
| XM_017010197.1 | XP_016865686.1 |
More Information
Set Membership: |
|

Genomic Map