Assay Details
Target Gene Details
Entrez Gene ID: | 644943 |
Gene Name: | Ras association domain family member 10 |
Gene Aliases: |
- |
Location: |
Chr.11:13009423-13012106 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASSF10 | NM_001080521.2 | 1 | 196 | NP_001073990.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3625402 | Chr.11:12851052 - 13035904 on Build GRCh38 | Gain |
|
| esv3625395 | Chr.11:12409149 - 13018985 on Build GRCh38 | Gain |
|
| nsv951309 | Chr.11:13008654 - 13011453 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map