Assay Details
Target Gene Details
Entrez Gene ID: | 145581 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 5 |
Gene Aliases: |
C14orf146, FIGLER8, SALM5 |
Location: |
Chr.14:41604684-41904549 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRFN5 | XM_005267369.3 | 1 | 890 | XP_005267426.1 |
| XM_017021050.1 | 1 | 890 | XP_016876539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759983 | Chr.14:41576670 - 41790308 on Build GRCh38 | Loss |
|
| nsv1042078 | Chr.14:41524782 - 41650940 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map