Assay Details
Target Gene Details
Entrez Gene ID: | 161176 |
Gene Name: | spectrin repeat containing nuclear envelope family member 3 |
Gene Aliases: |
C14orf49, NET53, Nesp3 |
Location: |
Chr.14:95416083-95516693 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNE3 | NM_152592.3 | NP_689805.3 | ||
| XM_005267377.3 | XP_005267434.1 | |||
| XM_006720063.3 | XP_006720126.1 | |||
| XM_011536515.2 | XP_011534817.1 | |||
| XM_017021056.1 | XP_016876545.1 | |||
| AK098471.1 | BAC05312.1 | |||
| AK131436.1 | BAD18582.1 | |||
| BC146604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635400 | Chr.14:95324080 - 95552963 on Build GRCh38 | Gain |
|
| nsv565631 | Chr.14:95320320 - 95666158 on Build GRCh38 | Gain |
|
| dgv89e55 | Chr.14:95292222 - 95675258 on Build GRCh38 | Gain |
|
| nsv1042858 | Chr.14:95326385 - 95660745 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map