Assay Details
Target Gene Details
Entrez Gene ID: | 3320 |
Gene Name: | heat shock protein 90 alpha family class A member 1 |
Gene Aliases: |
EL52, HEL-S-65p, HSP86, HSP89A, HSP90A, HSP90N, HSPC1, HSPCA, HSPCAL1, HSPCAL4, HSPN, Hsp89, Hsp90, LAP-2, LAP2 |
Location: |
Chr.14:102080738-102139749 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSP90AA1 | NM_001017963.2 | NP_001017963.2 | ||
| NM_005348.3 | NP_005339.3 | |||
| XM_011536718.2 | XP_011535020.1 | |||
| AF028832.1 | AAC25497.1 | |||
| AJ890082.1 | CAI64495.1 | |||
| AJ890083.1 | CAI64496.1 | |||
| AK056446.1 | ||||
| AK129557.1 | ||||
| AK291115.1 | ||||
| AK291607.1 | ||||
| AK300126.1 | ||||
| BC000987.2 | AAH00987.1 | |||
| BC007989.2 | AAH07989.2 | |||
| BC023006.1 | AAH23006.2 | |||
| BC108695.1 | AAI08696.1 | |||
| BC121062.2 | ||||
| BX247955.1 | CAD62296.1 | |||
| GQ149083.1 | ||||
| GQ472230.1 | ||||
| X15183.1 | CAA33259.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565947 | Chr.14:102001510 - 102204240 on Build GRCh38 | Gain |
|
| nsv1428 | Chr.14:102068860 - 102102356 on Build GRCh38 | Insertion |
|
| dgv25n68 | Chr.14:102000993 - 102195522 on Build GRCh38 | Loss |
|
| nsv983856 | Chr.14:102080748 - 102084595 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map