Assay Details
Target Gene Details
Entrez Gene ID: | 54550 |
Gene Name: | N-terminal EF-hand calcium binding protein 2 |
Gene Aliases: |
EFCBP2 |
Location: |
Chr.16:83968632-84002776 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECAB2 | NM_019065.2 | NP_061938.2 | ||
| XM_011523135.1 | XP_011521437.1 | |||
| XM_017023317.1 | XP_016878806.1 | |||
| AF070637.1 | AAC25392.1 | |||
| AF193758.1 | AAG28414.1 | |||
| AK127376.1 | BAC86948.1 | |||
| AY299331.1 | AAP57260.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3572242 | Chr.16:83975928 - 84007908 on Build GRCh38 | Gain |
|
| nsv528396 | Chr.16:83937637 - 84050522 on Build GRCh38 | Loss |
|
| nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss |
|
| esv3639405 | Chr.16:83969606 - 84018131 on Build GRCh38 | Loss |
|
| dgv5259n54 | Chr.16:83956484 - 84010961 on Build GRCh38 | Loss |
|
| dgv3056n100 | Chr.16:83923502 - 84039073 on Build GRCh38 | Loss |
|
| nsv471106 | Chr.16:83921780 - 84014793 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75244824] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map