Assay Details
Target Gene Details
Entrez Gene ID: | 59344 |
Gene Name: | arachidonate lipoxygenase 3 |
Gene Aliases: |
ARCI3, E-LOX, eLOX-3, eLOX3 |
Location: |
Chr.17:8095900-8118916 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALOXE3 | NM_001165960.1 | NP_001159432.1 | ||
| NM_021628.2 | NP_067641.2 | |||
| AF182218.1 | AAG16899.1 | |||
| AJ269499.1 | CAC12843.1 | |||
| AK292732.1 | ||||
| AK296416.1 | ||||
| AK313677.1 | ||||
| BC101938.1 | AAI01939.1 | |||
| BC101939.1 | AAI01940.1 | |||
| BC103508.1 | AAI03509.1 | |||
| BC104724.1 | AAI04725.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833352 | Chr.17:8029719 - 8187575 on Build GRCh38 | Loss |
|
| nsv574334 | Chr.17:8096271 - 8110990 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76084564] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map