Assay Details
Target Gene Details
Entrez Gene ID: | 84322 |
Gene Name: | chromosome 18 open reading frame 12 |
Gene Aliases: |
HEIL1, HsT2508 |
Location: |
Chr.18:48252301-48253185 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C18orf12 | XM_017026109.1 | 1 | 254 | XP_016881598.1 |
| AB027121.1 | 1 | 254 | BAB47121.1 |
Target Gene Details
Entrez Gene ID: | 201501 |
Gene Name: | zinc finger and BTB domain containing 7C |
Gene Aliases: |
APM-1, APM1, ZBTB36, ZNF857C |
Location: |
Chr.18:48026886-48412639 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
More Information
Set Membership: |
|

Genomic Map