Assay Details
Target Gene Details
Entrez Gene ID: | 10333 |
Gene Name: | toll like receptor 6 |
Gene Aliases: |
CD286 |
Location: |
Chr.4:38822897-38857767 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLR6 | NM_006068.4 | NP_006059.2 | ||
| XM_011513612.2 | XP_011511914.1 | |||
| XM_011513614.2 | XP_011511916.1 | |||
| AK313618.1 | ||||
| DA619469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3600267 | Chr.4:38809373 - 38862827 on Build GRCh38 | Loss |
|
| esv2672127 | Chr.4:38809351 - 38862791 on Build GRCh38 | Deletion |
|
| nsv818227 | Chr.4:38846296 - 38857139 on Build GRCh38 | Gain |
|
| nsv594010 | Chr.4:38804398 - 38873181 on Build GRCh38 | Loss |
|
| nsv950576 | Chr.4:38849480 - 38858279 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73811225] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map