Assay Details
Target Gene Details
Entrez Gene ID: | 2099 |
Gene Name: | estrogen receptor 1 |
Gene Aliases: |
ER, ESR, ESRA, ESTRR, Era, NR3A1 |
Location: |
Chr.6:151654148-152103274 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESR1 | XM_017010376.1 | 1 | 362 | XP_016865865.1 |
| XM_017010377.1 | 1 | 362 | XP_016865866.1 | |
| XM_017010380.1 | 1 | 362 | XP_016865869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830844 | Chr.6:151638649 - 151860891 on Build GRCh38 | Gain |
|
| esv2761004 | Chr.6:151528892 - 152050641 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map