Assay Details
Target Gene Details
Entrez Gene ID: | 137695 |
Gene Name: | transmembrane protein 68 |
Gene Aliases: |
- |
Location: |
Chr.8:55738744-55773407 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM68 | NM_001286657.1 | NP_001273586.1 | ||
| NM_001286661.1 | NP_001273590.1 | |||
| NM_152417.2 | NP_689630.1 | |||
| XM_005251150.1 | XP_005251207.1 | |||
| XM_011517454.1 | XP_011515756.1 | |||
| XM_011517455.2 | XP_011515757.1 | |||
| XM_017013040.1 | XP_016868529.1 | |||
| AK056932.1 | BAB71312.1 | |||
| AL832935.1 | CAH56335.1 | |||
| CN272366.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv459n21 | Chr.8:55676869 - 56030580 on Build GRCh38 | Gain |
|
| nsv527341 | Chr.8:55679732 - 55895699 on Build GRCh38 | Gain |
|
| esv2422388 | Chr.8:55707094 - 55812437 on Build GRCh38 | Duplication |
|
| nsv611392 | Chr.8:55741553 - 55878302 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map