Assay Details
Target Gene Details
Entrez Gene ID: | 286183 |
Gene Name: | Na+/K+ transporting ATPase interacting 3 |
Gene Aliases: |
FAM77D |
Location: |
Chr.8:62248942-62999652 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKAIN3 | NM_001304533.1 | NP_001291462.1 | ||
| NR_130764.1 | ||||
| XM_011517511.2 | XP_011515813.1 | |||
| XM_011517512.2 | XP_011515814.1 | |||
| XM_017013359.1 | XP_016868848.1 | |||
| XM_017013360.1 | XP_016868849.1 | |||
| AK096949.1 | BAC04909.1 | |||
| AK299689.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019143 | Chr.8:62229643 - 62478261 on Build GRCh38 | Loss |
|
| esv3891406 | Chr.8:61776965 - 62523810 on Build GRCh38 | Gain |
|
| esv2759621 | Chr.8:62243564 - 62347623 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map