Assay Details
Target Gene Details
Entrez Gene ID: | 128506 |
Gene Name: | osteoclast stimulatory transmembrane protein |
Gene Aliases: |
C20orf123, OC-STAMP, dJ257E24.3 |
Location: |
Chr.20:46541031-46550574 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OCSTAMP | NM_080721.2 | NP_542452.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833994 | Chr.20:46442116 - 46680210 on Build GRCh38 | Gain |
|
| nsv1061899 | Chr.20:46498425 - 47439490 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map