Assay Details
Target Gene Details
Entrez Gene ID: | 8242 |
Gene Name: | lysine demethylase 5C |
Gene Aliases: |
DXS1272E, JARID1C, MRX13, MRXJ, MRXSCJ, MRXSJ, SMCX, XE169 |
Location: |
Chr.X:53159735-53225422 on Build GRCh38 |
Assay Gene Location: | Within Intron 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM5C | XM_011530827.2 | XP_011529129.1 | ||
| XM_017029886.1 | XP_016885375.1 | |||
| XM_017029887.1 | XP_016885376.1 | |||
| XM_017029888.1 | XP_016885377.1 | |||
| EF613277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map