Assay Details
Target Gene Details
Entrez Gene ID: | 58190 |
Gene Name: | CTD small phosphatase 1 |
Gene Aliases: |
NIF3, NLI-IF, NLIIF, SCP1 |
Location: |
Chr.2:218398338-218405941 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTDSP1 | NM_001206878.1 | NP_001193807.1 | ||
| XM_017004617.1 | 1 | 284 | XP_016860106.1 | |
| BX424971.2 | ||||
| BX446444.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519829 | Chr.2:218286495 - 218403058 on Build GRCh38 | Loss |
|
| nsv3166 | Chr.2:218371765 - 218405183 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map