Assay Details
Target Gene Details
Entrez Gene ID: | 55843 |
Gene Name: | Rho GTPase activating protein 15 |
Gene Aliases: |
BM046 |
Location: |
Chr.2:143070973-143768352 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP15 | XM_011511479.2 | XP_011509781.1 | ||
| XM_017004499.1 | XP_016859988.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2953 | Chr.2:143054294 - 143085719 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map