Assay Details
Target Gene Details
Entrez Gene ID: | 29842 |
Gene Name: | transcription factor CP2-like 1 |
Gene Aliases: |
CRTR1, LBP-9, LBP9 |
Location: |
Chr.2:121216587-121285210 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFCP2L1 | NM_014553.2 | 15 | 9163 | NP_055368.1 |
| XM_017003902.1 | 14 | 9028 | XP_016859391.1 | |
| XM_017003903.1 | 10 | 8470 | XP_016859392.1 | |
| BM055156.1 | 1 | 297 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582798 | Chr.2:120552693 - 121587495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map