Assay Details
Target Gene Details
Entrez Gene ID: | 6326 |
Gene Name: | sodium voltage-gated channel alpha subunit 2 |
Gene Aliases: |
BFIC3, BFIS3, BFNIS, EIEE11, HBA, HBSCI, HBSCII, NAC2, Na(v)1.2, Nav1.2, SCN2A1, SCN2A2 |
Location: |
Chr.2:165208056-165392310 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN2A | NM_001040142.1 | NP_001035232.1 | ||
| XM_005246753.3 | XP_005246810.1 | |||
| XM_017004656.1 | XP_016860145.1 | |||
| XM_017004658.1 | XP_016860147.1 | |||
| AK289656.1 | ||||
| BP199510.1 |
More Information
Set Membership: |
|

Genomic Map