Assay Details
Target Gene Details
Entrez Gene ID: | 150468 |
Gene Name: | cytoskeleton associated protein 2 like |
Gene Aliases: |
- |
Location: |
Chr.2:112736349-112764677 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CKAP2L | NM_001304361.1 | NP_001291290.1 | ||
| NM_152515.4 | NP_689728.3 | |||
| NR_130712.1 | ||||
| XM_011510666.2 | XP_011508968.1 | |||
| AK097948.1 | BAC05202.1 | |||
| AK292530.1 | ||||
| AK302875.1 | ||||
| AL832036.1 | ||||
| BC036217.1 | AAH36217.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs72952109] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map