Assay Details
Target Gene Details
Entrez Gene ID: | 776 |
Gene Name: | calcium voltage-gated channel subunit alpha1 D |
Gene Aliases: |
CACH3, CACN4, CACNL1A2, CCHL1A2, Cav1.3, PASNA, SANDD |
Location: |
Chr.3:53495049-53813151 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA1D | NM_000720.3 | NP_000711.1 | ||
| NM_001128839.2 | NP_001122311.1 | |||
| NM_001128840.2 | NP_001122312.1 | |||
| XM_005265448.3 | XP_005265505.1 | |||
| XM_011534094.2 | XP_011532396.1 | |||
| XM_011534096.2 | XP_011532398.1 | |||
| XM_011534097.2 | XP_011532399.1 | |||
| XM_011534100.2 | XP_011532402.1 | |||
| XM_017007137.1 | XP_016862626.1 | |||
| XM_017007138.1 | XP_016862627.1 | |||
| XM_017007139.1 | XP_016862628.1 | |||
| XM_017007140.1 | XP_016862629.1 | |||
| XM_017007141.1 | XP_016862630.1 | |||
| XM_017007142.1 | XP_016862631.1 | |||
| XM_017007143.1 | XP_016862632.1 | |||
| XM_017007144.1 | XP_016862633.1 | |||
| XM_017007145.1 | XP_016862634.1 | |||
| EU363339.1 | ||||
| M76558.1 | AAA58402.1 | |||
| M83566.1 | AAA35629.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2725284 | Chr.3:53517229 - 53517337 on Build GRCh38 | Deletion |
|
| nsv1003621 | Chr.3:53371619 - 54391295 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map