Assay Details
Target Gene Details
Entrez Gene ID: | 201895 |
Gene Name: | small integral membrane protein 14 |
Gene Aliases: |
C4orf34 |
Location: |
Chr.4:39547297-39638998 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMIM14 | NM_001317896.1 | NP_001304825.1 | ||
| NM_001317897.1 | NP_001304826.1 | |||
| NM_174921.2 | NP_777581.1 | |||
| XM_011513640.2 | XP_011511942.1 | |||
| XM_017007862.1 | XP_016863351.1 | |||
| XM_017007863.1 | XP_016863352.1 | |||
| AK023351.1 | ||||
| AK307146.1 | ||||
| BC008502.1 | AAH08502.1 | |||
| BQ049060.1 | ||||
| HY004916.1 | ||||
| HY155982.1 |
Target Gene Details
Entrez Gene ID: | 100885776 |
Gene Name: | UGDH antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:39527839-39594707 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGDH-AS1 | NR_047679.1 | |||
| AK056558.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997937 | Chr.4:39568834 - 39635710 on Build GRCh38 | Gain |
|

Genomic Map