Assay Details
Target Gene Details
Entrez Gene ID: | 90826 |
Gene Name: | protein arginine methyltransferase 9 |
Gene Aliases: |
PRMT10 |
Location: |
Chr.4:147635626-147684230 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRMT9 | NM_001304458.1 | NP_001291387.1 | ||
| NM_138364.3 | NP_612373.2 | |||
| XM_017008823.1 | XP_016864312.1 | |||
| XM_017008824.1 | XP_016864313.1 | |||
| XM_017008825.1 | XP_016864314.1 | |||
| XM_017008826.1 | XP_016864315.1 | |||
| AK096703.1 | ||||
| AK292904.1 | ||||
| BC021250.2 | AAH21250.2 | |||
| BC064403.1 | AAH64403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830110 | Chr.4:147510973 - 147673968 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map