Assay Details
Target Gene Details
Entrez Gene ID: | 105379393 |
Gene Name: | uncharacterized LOC105379393 |
Gene Aliases: |
- |
Location: |
Chr.8:42051750-42104987 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379393 | NR_136589.1 | |||
| HY030269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv397995 | Chr.8:42053634 - 42054262 on Build GRCh38 | Deletion |
|
| nsv521189 | Chr.8:42050630 - 42056486 on Build GRCh38 | Loss |
|
| nsv824615 | Chr.8:42049114 - 42054345 on Build GRCh38 | Gain |
|

Genomic Map