Assay Details
Target Gene Details
Entrez Gene ID: | 56165 |
Gene Name: | tudor domain containing 1 |
Gene Aliases: |
CT41.1 |
Location: |
Chr.10:114174442-114232669 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDRD1 | NM_198795.1 | NP_942090.1 | ||
| XM_005269978.2 | XP_005270035.1 | |||
| XM_005269979.2 | XP_005270036.1 | |||
| XM_011539959.2 | XP_011538261.1 | |||
| XM_011539960.2 | XP_011538262.1 | |||
| XM_011539961.2 | XP_011538263.1 | |||
| XM_011539962.1 | XP_011538264.1 | |||
| XM_011539963.1 | XP_011538265.1 | |||
| XM_011539964.1 | XP_011538266.1 | |||
| XM_011539966.1 | XP_011538268.1 | |||
| XM_017016414.1 | XP_016871903.1 | |||
| XM_017016415.1 | XP_016871904.1 | |||
| AK122916.1 | ||||
| AL833340.1 | ||||
| BC035010.2 | AAH35010.1 | |||
| BC063133.1 | AAH63133.1 | |||
| BX648664.1 |
More Information
Set Membership: |
|

Genomic Map