Assay Details
Target Gene Details
Entrez Gene ID: | 5464 |
Gene Name: | pyrophosphatase (inorganic) 1 |
Gene Aliases: |
HEL-S-66p, IOPPP, PP, PP1, SID6-8061 |
Location: |
Chr.10:70202830-70233434 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPA1 | NM_021129.3 | NP_066952.1 | ||
| AB026723.1 | BAA84702.1 | |||
| AF092439.1 | AAP97214.1 | |||
| AF108211.1 | AAD24964.1 | |||
| AF119665.1 | AAF17222.1 | |||
| AF154065.1 | AAD34643.1 | |||
| AF217186.1 | AAG36780.1 | |||
| AK058193.1 | ||||
| BC001022.2 | ||||
| BC061581.1 | ||||
| BC105034.1 | AAI05035.1 | |||
| BC105036.1 | AAI05037.1 | |||
| BC107882.1 | AAI07883.1 | |||
| BU520836.1 | ||||
| EU794626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss |
|
| nsv831908 | Chr.10:70210726 - 70389945 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80272958] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map