Assay Details
Target Gene Details
Entrez Gene ID: | 64421 |
Gene Name: | DNA cross-link repair 1C |
Gene Aliases: |
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C |
Location: |
Chr.10:14897853-14954432 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCLRE1C | NM_001033855.2 | 14 | 5425 | NP_001029027.1 |
| NM_001033857.2 | 15 | 5510 | NP_001029029.1 | |
| NM_001033858.2 | 16 | 5832 | NP_001029030.1 | |
| NM_001289076.1 | 12 | 5284 | NP_001276005.1 | |
| NM_001289077.1 | 15 | 5556 | NP_001276006.1 | |
| NM_001289078.1 | 12 | 5317 | NP_001276007.1 | |
| NM_001289079.1 | 16 | 5878 | NP_001276008.1 | |
| NM_022487.3 | 13 | 5369 | NP_071932.2 | |
| NR_110297.1 | 17 | 5778 | ||
| XM_017016556.1 | XP_016872045.1 | |||
| XM_017016559.1 | XP_016872048.1 | |||
| XM_017016560.1 | XP_016872049.1 | |||
| BC022254.1 | AAH22254.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469720 | Chr.10:14865281 - 15037379 on Build GRCh38 | Gain |
|
| nsv831793 | Chr.10:14870604 - 15058763 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map