Assay Details
Target Gene Details
Entrez Gene ID: | 2585 |
Gene Name: | galactokinase 2 |
Gene Aliases: |
GK2 |
Location: |
Chr.15:49155757-49367883 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALK2 | NM_001001556.2 | NP_001001556.1 | ||
| XM_005254280.3 | XP_005254337.1 | |||
| XM_005254284.3 | XP_005254341.1 | |||
| XM_017022063.1 | XP_016877552.1 | |||
| XM_017022065.1 | XP_016877554.1 | |||
| AF461816.1 | AAP97708.1 | |||
| AL832745.1 | ||||
| DC390737.1 |
Target Gene Details
Entrez Gene ID: | 100616332 |
Gene Name: | microRNA 4716 |
Gene Aliases: |
mir-4716 |
Location: |
Chr.15:49169070-49169153 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4716 | NR_039866.1 |
More Information
Set Membership: |
|

Genomic Map