Assay Details
Target Gene Details
Entrez Gene ID: | 10239 |
Gene Name: | adaptor related protein complex 3 sigma 2 subunit |
Gene Aliases: |
AP3S3, sigma3b |
Location: |
Chr.15:89830599-89894385 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP3S2 | NM_005829.4 | NP_005820.1 | ||
| NR_023361.1 | ||||
| NR_037582.1 | ||||
| AK222698.1 | BAD96418.1 | |||
| AK294712.1 | ||||
| AK312467.1 | ||||
| AL110108.1 | ||||
| BC002785.2 | AAH02785.1 | |||
| BC007773.2 | AAH07773.1 | |||
| BC010020.2 | AAH10020.1 | |||
| DA777624.1 | ||||
| DC340463.1 | ||||
| X99459.1 | CAA67824.1 |
Target Gene Details
Entrez Gene ID: | 100526783 |
Gene Name: | C15orf38-AP3S2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.15:89830599-89912990 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C15orf38-AP3S2 | NM_001199058.1 | NP_001185987.1 | ||
| AK000495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2667255 | Chr.15:89846945 - 89851628 on Build GRCh38 | Deletion |
|
| esv3637203 | Chr.15:89845033 - 89856267 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map