This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05364297_cn
See other HACD3 CNV Assays ›
Gene Symbol
HACD3
Assay Reference Genome
Location

Chr.15:65532950 on build GRCh38
Cytoband
15q22.31
Assay ID Hs05364297_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

51495

Gene Name:

3-hydroxyacyl-CoA dehydratase 3

Gene Aliases:

B-IND1, BIND1, HSPC121, PTPLAD1

Location:

Chr.15:65530489-65578355 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
HACD3 NM_016395.2 NP_057479.2
XM_011521664.1 XP_011519966.1
AF161470.1 AAF29085.1
AJ271091.1 CAB69070.1
AK027421.1 BAB55101.1
AK074898.1 BAC11277.1
AK225120.1
AK299234.1
BC035508.1 AAH35508.1
BC047685.1 AAH47685.1
BC058912.1 AAH58912.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833042 Chr.15:65446079 - 65656395 on Build GRCh38 Loss SLC24A1 HACD3 DPP8 VWA9
nsv569787 Chr.15:65525552 - 65538180 on Build GRCh38 Loss HACD3

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

I-kappaB kinase/NF-kappaB signaling
activation of JUN kinase activity
small GTPase mediated signal transduction
Rho protein signal transduction
Rac protein signal transduction
fatty acid elongation
very long-chain fatty acid biosynthetic process
positive regulation of GTPase activity
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
GTPase activator activity
protein binding
3-hydroxyacyl-CoA dehydratase activity
enzyme binding
3-hydroxy-behenoyl-CoA dehydratase activity
3-hydroxy-lignoceroyl-CoA dehydratase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6479c7f679-sz4vp:80/100.66.76.236:80.
git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.50.0-Offline