Assay Details
Target Gene Details
Entrez Gene ID: | 54839 |
Gene Name: | leucine rich repeat containing 49 |
Gene Aliases: |
PGs4 |
Location: |
Chr.15:70852671-71050097 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC49 | NM_001284357.1 | NP_001271286.1 | ||
| XM_017022357.1 | XP_016877846.1 | |||
| XM_017022359.1 | XP_016877848.1 | |||
| AK123672.1 | ||||
| DA395472.1 |
More Information
Set Membership: |
|

Genomic Map