Assay Details
Target Gene Details
Entrez Gene ID: | 150082 |
Gene Name: | LCA5L, lebercilin like |
Gene Aliases: |
C21orf13 |
Location: |
Chr.21:39405844-39445796 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647018 | Chr.21:39406469 - 39425776 on Build GRCh38 | Loss |
|
| dgv4433n100 | Chr.21:39369967 - 39418271 on Build GRCh38 | Gain |
|
| nsv1061454 | Chr.21:39386047 - 39893479 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map