Assay Details
Target Gene Details
Entrez Gene ID: | 158 |
Gene Name: | adenylosuccinate lyase |
Gene Aliases: |
AMPS, ASASE, ASL |
Location: |
Chr.22:40346500-40387408 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADSL | XM_011529979.1 | XP_011528281.1 |
Target Gene Details
Entrez Gene ID: | 27352 |
Gene Name: | small G protein signaling modulator 3 |
Gene Aliases: |
MAP, RABGAP5, RUSC3, RUTBC3, RabGAP-5, rabGAPLP |
Location: |
Chr.22:40370530-40410289 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834202 | Chr.22:40179762 - 40385241 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map