Assay Details
Target Gene Details
Entrez Gene ID: | 414918 |
Gene Name: | DENN domain containing 6B |
Gene Aliases: |
AFI1B, FAM116B |
Location: |
Chr.22:50309032-50327088 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND6B | XM_011530692.2 | 21 | 4460 | XP_011528994.1 |
| XM_011530693.2 | 20 | 4239 | XP_011528995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955199 | Chr.22:50149372 - 50320271 on Build GRCh38 | Deletion |
|
| dgv781n67 | Chr.22:50153213 - 50314433 on Build GRCh38 | Gain |
|
| nsv589209 | Chr.22:50284572 - 50315934 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map