Assay Details
Target Gene Details
Entrez Gene ID: | 54885 |
Gene Name: | TBC1 domain family member 8B |
Gene Aliases: |
GRAMD8B |
Location: |
Chr.X:106802689-106876150 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D8B | NM_017752.2 | NP_060222.2 | ||
| NM_198881.1 | NP_942582.1 | |||
| XM_006724669.1 | XP_006724732.1 | |||
| XM_006724670.3 | XP_006724733.1 | |||
| AB449890.1 | ||||
| AB449891.1 | ||||
| AK123957.1 | BAC85735.1 | |||
| BC122564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752312 | Chr.X:105705935 - 107123872 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73531165] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map