Assay Details
Target Gene Details
Entrez Gene ID: | 115350 |
Gene Name: | Fc receptor like 1 |
Gene Aliases: |
CD307a, FCRH1, IFGP1, IRTA5 |
Location: |
Chr.1:157792249-157820150 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
More Information
Set Membership: |
|

Genomic Map