Assay Details
Target Gene Details
Entrez Gene ID: | 81627 |
Gene Name: | tRNA methyltransferase 1 like |
Gene Aliases: |
C1orf25, MST070, MSTP070, TRM1L, bG120K12.3 |
Location: |
Chr.1:185118085-185157098 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT1L | NM_001202423.1 | NP_001189352.1 | ||
| NM_030934.4 | NP_112196.3 | |||
| XM_011510028.2 | XP_011508330.1 | |||
| AF163261.1 | AAQ13590.1 | |||
| AF288399.1 | AAG60618.1 | |||
| AK074936.1 | BAC11302.1 | |||
| AK074993.1 | BAC11341.1 | |||
| AK098359.1 | ||||
| AK131460.1 | BAD18605.1 | |||
| AK226103.1 | ||||
| AK302166.1 | ||||
| AK312801.1 | ||||
| BC045535.1 | AAH45535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548396 | Chr.1:185068972 - 185182246 on Build GRCh38 | Loss |
|
| nsv521613 | Chr.1:185102699 - 185202737 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map