Assay Details
Target Gene Details
Entrez Gene ID: | 11343 |
Gene Name: | monoglyceride lipase |
Gene Aliases: |
HU-K5, HUK5, MAGL, MGL |
Location: |
Chr.3:127689062-127823250 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGLL | NM_001003794.2 | NP_001003794.1 | ||
| NM_001256585.1 | NP_001243514.1 | |||
| NM_007283.6 | NP_009214.1 | |||
| XM_011512377.1 | XP_011510679.1 | |||
| XM_011512378.1 | XP_011510680.1 | |||
| XM_011512379.2 | XP_011510681.1 | |||
| XM_011512382.1 | XP_011510684.1 | |||
| XM_011512383.2 | XP_011510685.1 | |||
| XM_017005664.1 | XP_016861153.1 | |||
| XM_017005665.1 | XP_016861154.1 | |||
| XM_017005666.1 | XP_016861155.1 | |||
| AI125141.1 | ||||
| AJ270950.1 | CAC43316.1 | |||
| AK091314.1 | ||||
| AK304844.1 | ||||
| AK315529.1 | ||||
| AL365376.1 | ||||
| AY172950.1 | AAO45694.1 | |||
| BC000551.2 | AAH00551.1 | |||
| BC006230.2 | AAH06230.1 | |||
| BG035493.1 | ||||
| BM669411.1 | ||||
| BX403703.2 | ||||
| BX640777.1 | CAE45873.1 | |||
| CR456835.1 | CAG33116.1 | |||
| EU660904.1 | ||||
| U67963.1 | AAB39616.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460848 | Chr.3:127667659 - 127702011 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map