Assay Details
Target Gene Details
Entrez Gene ID: | 6386 |
Gene Name: | syndecan binding protein |
Gene Aliases: |
MDA-9, MDA9, ST1, SYCL, TACIP18 |
Location: |
Chr.8:58553169-58582860 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDCBP | NM_001007067.1 | NP_001007068.1 | ||
| NM_001007068.1 | NP_001007069.1 | |||
| NM_001007069.1 | NP_001007070.1 | |||
| NM_001007070.1 | NP_001007071.1 | |||
| NM_005625.3 | NP_005616.2 | |||
| AF000652.1 | AAB97144.1 | |||
| AF006636.1 | AAC52050.1 | |||
| AK128645.1 | ||||
| AK295198.1 | ||||
| AK298697.1 | ||||
| AK300647.1 | ||||
| AK312274.1 | ||||
| BC113674.1 | ||||
| BC113676.1 | ||||
| BC143915.1 | ||||
| BC143916.1 | ||||
| BG772269.1 |
More Information
Set Membership: |
|

Genomic Map