Assay Details
Target Gene Details
Entrez Gene ID: | 100532726 |
Gene Name: | NDUFC2-KCTD14 readthrough |
Gene Aliases: |
- |
Location: |
Chr.11:78015715-78080219 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFC2-KCTD14 | NM_001203260.1 | NP_001190189.1 | ||
| NM_001203261.1 | NP_001190190.1 | |||
| NM_001203262.1 | NP_001190191.1 | |||
| BG723290.1 | ||||
| FY211285.1 | ||||
| FY212410.1 | ||||
| FY212967.1 | ||||
| FY213449.1 |
Target Gene Details
Entrez Gene ID: | 7069 |
Gene Name: | thyroid hormone responsive |
Gene Aliases: |
LPGP1, Lpgp, S14, SPOT14, THRP |
Location: |
Chr.11:78063861-78068357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THRSP | NM_003251.3 | NP_003242.1 | ||
| AI480357.1 | ||||
| BC031989.1 | AAH31989.1 | |||
| BG674197.1 | ||||
| BP317892.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain |
|
| nsv1050891 | Chr.11:77705600 - 78076956 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map