Assay Details
Target Gene Details
Entrez Gene ID: | 56649 |
Gene Name: | transmembrane protease, serine 4 |
Gene Aliases: |
CAPH2, MT-SP2, TMPRSS3 |
Location: |
Chr.11:118077012-118125505 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Gene Details
Entrez Gene ID: | 100526771 |
Gene Name: | TMPRSS4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:118015772-118086793 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS4-AS1 | NR_038318.1 | |||
| AB231754.1 | ||||
| AB231755.1 | ||||
| AB231756.1 | ||||
| AB231757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468878 | Chr.11:118076983 - 118161527 on Build GRCh38 | Gain |
|
| nsv1049373 | Chr.11:118066351 - 118134273 on Build GRCh38 | Loss |
|
| nsv556463 | Chr.11:118061915 - 118099611 on Build GRCh38 | Loss |
|
| nsv1159938 | Chr.11:118075976 - 118157452 on Build GRCh38 | Duplication |
|
| nsv509444 | Chr.11:118033438 - 118117536 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map