Assay Details
Target Gene Details
Entrez Gene ID: | 5936 |
Gene Name: | RNA binding motif protein 4 |
Gene Aliases: |
LARK, RBM4A, ZCCHC21, ZCRB3A |
Location: |
Chr.11:66638617-66668385 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM4 | NM_001198844.1 | NP_001185773.1 | ||
| AK303984.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518919 | Chr.11:66641938 - 66700998 on Build GRCh38 | Loss |
|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116732250] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map