Assay Details
Target Gene Details
Entrez Gene ID: | 27087 |
Gene Name: | beta-1,3-glucuronyltransferase 1 |
Gene Aliases: |
CD57, GLCATP, GLCUATP, HNK1, LEU7, NK-1, NK1 |
Location: |
Chr.11:134378504-134411986 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3GAT1 | NM_018644.3 | NP_061114.2 | ||
| NM_054025.2 | NP_473366.1 | |||
| XM_005271506.3 | XP_005271563.1 | |||
| XM_011542751.2 | XP_011541053.1 | |||
| XM_011542753.2 | XP_011541055.1 | |||
| XM_017017551.1 | XP_016873040.1 | |||
| AB029396.1 | BAA96077.1 | |||
| BC010466.1 | AAH10466.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556608 | Chr.11:134348406 - 134876236 on Build GRCh38 | Gain |
|
| nsv521581 | Chr.11:134126584 - 134772738 on Build GRCh38 | Gain |
|
| dgv1306n100 | Chr.11:134373155 - 134500539 on Build GRCh38 | Gain |
|
| dgv2206n54 | Chr.11:134381883 - 134555421 on Build GRCh38 | Gain |
|
| dgv4e196 | Chr.11:134304439 - 134744665 on Build GRCh38 | Duplication |
|
| dgv1305n100 | Chr.11:134363845 - 134416469 on Build GRCh38 | Gain |
|
| nsv556611 | Chr.11:134390175 - 134412392 on Build GRCh38 | Loss |
|
| esv3628169 | Chr.11:134353064 - 134485281 on Build GRCh38 | Gain |
|
| dgv1304n100 | Chr.11:134322594 - 134754120 on Build GRCh38 | Gain |
|
| esv2760555 | Chr.11:134282136 - 134860776 on Build GRCh38 | Gain+Loss |
|
| dgv1307n100 | Chr.11:134388248 - 134547865 on Build GRCh38 | Gain |
|
| esv3628170 | Chr.11:134357170 - 134435562 on Build GRCh38 | Gain |
|
| nsv521623 | Chr.11:134307381 - 134756993 on Build GRCh38 | Gain |
|
| nsv1042300 | Chr.11:134386590 - 134414596 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78975867] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map