Assay Details
Target Gene Details
Entrez Gene ID: | 219865 |
Gene Name: | olfactory receptor family 8 subfamily G member 5 |
Gene Aliases: |
OR11-298, OR8G5P, OR8G6 |
Location: |
Chr.11:124241158-124323283 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR8G5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826113 | Chr.11:124267376 - 124372263 on Build GRCh38 | Loss |
|
| dgv82n21 | Chr.11:123901052 - 124289279 on Build GRCh38 | Gain |
|

Genomic Map