This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06326798_cn
See other AAMDC CNV Assays ›
Gene Symbol
AAMDC
Assay Reference Genome
Location

Chr.11:77822553 on build GRCh38
Cytoband
11q14.1
Assay ID Hs06326798_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

28971

Gene Name:

adipogenesis associated Mth938 domain containing

Gene Aliases:

C11orf67, CK067, PTD015

Location:

Chr.11:77821114-77901281 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
AAMDC NM_001316957.1 NP_001303886.1
NM_001316958.1 NP_001303887.1
NM_001316960.1 NP_001303889.1
NM_001316961.1 NP_001303890.1
NM_001316962.1 NP_001303891.1
NM_024684.3 NP_078960.1
NR_133637.1
XM_011544969.2 XP_011543271.1
AF092136.1 AAD40378.1
AI569731.1
AK024688.1 BAB14963.1
BC002752.2 AAH02752.1
BC016854.2 AAH16854.1
BI791786.1
BM930331.1
BP270361.1
BX646013.1
CN362204.1

Target Gene Details

Entrez Gene ID:

51773

Gene Name:

remodeling and spacing factor 1

Gene Aliases:

HBXAP, RSF-1, XAP8, p325

Location:

Chr.11:77660009-77872232 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
RSF1 XM_017017923.1 XP_016873412.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1050891 Chr.11:77705600 - 78076956 on Build GRCh38 Gain NDUFC2 KCTD14 RPS20P27 NDUFC2-KCTD14 RSF1 AAMDC THRSP INTS4

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of apoptotic process
positive regulation of fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
nucleosome assembly
chromatin remodeling
DNA-templated transcription, initiation
covalent chromatin modification
nucleosome positioning
CENP-A containing nucleosome assembly
negative regulation of DNA binding
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of viral transcription
protein binding
zinc ion binding
ATPase activity
histone binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline