Assay Details
Target Gene Details
Entrez Gene ID: | 8715 |
Gene Name: | nucleolar protein 4 |
Gene Aliases: |
CT125, HRIHFB2255, NOLP |
Location: |
Chr.18:33851100-34224834 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2668525 | Chr.18:33248704 - 33887857 on Build GRCh38 | Deletion |
|
| esv3642176 | Chr.18:33873158 - 33876520 on Build GRCh38 | Loss |
|
| nsv458048 | Chr.18:33840046 - 33917382 on Build GRCh38 | Loss |
|
| nsv576643 | Chr.18:33671257 - 34001283 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map