Assay Details
Target Gene Details
Entrez Gene ID: | 10390 |
Gene Name: | choline/ethanolamine phosphotransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.1:111139436-111185102 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEPT1 | NM_001007794.1 | NP_001007795.1 | ||
| NM_006090.3 | NP_006081.1 | |||
| XM_006710285.3 | XP_006710348.1 | |||
| XM_011540496.1 | XP_011538798.1 | |||
| AF068302.1 | AAD25170.1 | |||
| AK023381.1 | ||||
| AL833102.1 | CAH10403.1 | |||
| BC032610.2 | AAH32610.1 | |||
| BC049196.1 | AAH49196.1 | |||
| BX487092.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10639 | Chr.1:111126227 - 111244431 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map