Assay Details
Target Gene Details
Entrez Gene ID: | 10753 |
Gene Name: | calpain 9 |
Gene Aliases: |
GC36, nCL-4 |
Location: |
Chr.1:230747289-230803356 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPN9 | NM_001319676.1 | NP_001306605.1 | ||
| NM_006615.2 | NP_006606.1 | |||
| NM_016452.2 | NP_057536.1 | |||
| XM_011544017.1 | XP_011542319.1 | |||
| XM_011544018.1 | XP_011542320.1 | |||
| XM_011544019.2 | XP_011542321.1 | |||
| XM_011544020.1 | XP_011542322.1 | |||
| XM_017000098.1 | XP_016855587.1 | |||
| XM_017000099.1 | XP_016855588.1 | |||
| AB038463.1 | BAA92137.1 | |||
| AF022799.1 | AAB80762.1 | |||
| AK298024.1 | ||||
| AK309039.1 | ||||
| AK313629.1 | ||||
| BC027993.1 | AAH27993.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain |
|
| nsv549335 | Chr.1:230733804 - 230797361 on Build GRCh38 | Gain |
|
| nsv832836 | Chr.1:230693328 - 230832263 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map