Assay Details
Target Gene Details
Entrez Gene ID: | 55223 |
Gene Name: | tripartite motif containing 62 |
Gene Aliases: |
DEAR1 |
Location: |
Chr.1:33144603-33184774 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM62 | NM_018207.2 | NP_060677.2 | ||
| XM_011541705.2 | XP_011540007.1 | |||
| XM_017001629.1 | XP_016857118.1 | |||
| XM_017001630.1 | XP_016857119.1 | |||
| XM_017001631.1 | XP_016857120.1 | |||
| AK001621.1 | BAA91792.1 | |||
| AK122896.1 | ||||
| AK300177.1 | ||||
| AX775831.1 | CAE11615.1 | |||
| BC001222.1 | AAH01222.1 | |||
| BC007999.1 | AAH07999.1 | |||
| BC011689.2 | AAH11689.1 | |||
| BC012152.1 | AAH12152.1 |
More Information
Set Membership: |
|

Genomic Map