Assay Details
Target Gene Details
Entrez Gene ID: | 63924 |
Gene Name: | cell death inducing DFFA like effector c |
Gene Aliases: |
CIDE-3, CIDE3, FPLD5, FSP27 |
Location: |
Chr.3:9866710-9880254 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIDEC | NM_001199551.1 | NP_001186480.1 | ||
| NM_001199552.1 | NP_001186481.1 | |||
| NM_001321142.1 | NP_001308071.1 | |||
| NM_001321143.1 | NP_001308072.1 | |||
| NM_001321144.1 | NP_001308073.1 | |||
| AK024524.1 | BAB14920.1 | |||
| AK024530.1 | BAB14922.1 | |||
| AL598731.1 | ||||
| BQ695178.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3595255 | Chr.3:9879400 - 9888341 on Build GRCh38 | Loss |
|
| esv2724890 | Chr.3:9879501 - 9888329 on Build GRCh38 | Deletion |
|
| nsv821983 | Chr.3:9879369 - 9887694 on Build GRCh38 | Loss |
|
| nsv834615 | Chr.3:9791053 - 9971639 on Build GRCh38 | Loss |
|
| esv2666111 | Chr.3:9879405 - 9888403 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map